Methyl 2-bromo-6-(trifluoromethyl)benzoate

Supplier: Apollo Scientific

PC502702-1G
APOSPC502702-1GEA 199 EUR
APOSPC502702-1G
Methyl 2-bromo-6-(trifluoromethyl)benzoate
Methyl 2-bromo-6-(trifluoromethyl)benzoate
Formula: C₉H₆BrF₃O₂
MW: 283.042 g/mol
Storage Temperature: Ambient
MDL Number: MFCD14697996
CAS Number: 1214324-11-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR