7-(4-Bromo-2-fluorophenyl)-5,8-diazaspiro[3.4]octan-6-one

Supplier: Apollo Scientific

PC510035-5G
APOSPC510035-5GEA 1220 EUR
APOSPC510035-5G
7-(4-Bromo-2-fluorophenyl)-5,8-diazaspiro[3.4]octan-6-one
7-(4-Bromo-2-fluorophenyl)-5,8-diazaspiro[3.4]octan-6-one
Formula: C₁₂H₁₂BrFN₂O
MW: 299.139 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18651746
CAS Number: 1272755-81-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR