2-(Allyloxy)-4,6-difluorobenzyl Alcohol

Supplier: Apollo Scientific

PC510149-5G
APOSPC510149-5GEA 916 EUR
APOSPC510149-5G
2-(Allyloxy)-4,6-difluorobenzyl Alcohol
2-(Allyloxy)-4,6-difluorobenzyl Alcohol
Formula: C₁₀H₁₀F₂O₂
MW: 200.182 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26937566
CAS Number: 1823900-28-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR