Fluoromalonaldehyde, sodium salt

Supplier: Apollo Scientific
Warning

Synonyms: Sodium 2-fluoro-3-oxoprop-1-en-1-olate, Fluoropropane-1,3-dial sodium salt

PC51104-1G PC51104-250MG
APOSPC51104-1GEA 319 EUR
APOSPC51104-1G APOSPC51104-250MG
Fluoromalonaldehyde, sodium salt
Fluoromalonaldehyde, sodium salt
Formula: C₃H₂FO₂·Na
MW: 112.035 g/mol
Melting Pt: >200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD21362228
CAS Number: 29548-72-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR