3-Fluoro-5-hydroxybenzaldehyde

Supplier: Apollo Scientific
Warning

Synonyms: 3-Fluoro-5-formylphenol

PC52056-1G PC52056-5G PC52056-10G
APOSPC52056-1GEA 29 EUR
APOSPC52056-1G APOSPC52056-5G APOSPC52056-10G
3-Fluoro-5-hydroxybenzaldehyde
3-Fluoro-5-hydroxybenzaldehyde
Formula: C₇H₅FO₂
MW: 140.112 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185579
CAS Number: 1023290-12-1

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR