Ethyl 2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-2,2-difluoroacetate

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 2-(3-chloro-5-(trifluoromethyl)pyridin-2-yl)-2,2-difluoroacetate

PC52170-1G
APOSPC52170-1GEA 237 EUR
APOSPC52170-1G
Ethyl 2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-2,2-difluoroacetate
Ethyl 2-[3-chloro-5-(trifluoromethyl)pyridin-2-yl]-2,2-difluoroacetate
Formula: C₁₀H₇ClF₅NO₂
MW: 303.613 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12546142
CAS Number: 1431842-79-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR