2-Chloro-3-(trifluoromethyl)benzaldehyde 98%

Supplier: Apollo Scientific
Warning

PC5289-500MG PC5289-5G
APOSPC5289-500MGEA 21.9 EUR
APOSPC5289-500MG APOSPC5289-5G
2-Chloro-3-(trifluoromethyl)benzaldehyde 98%
2-Chloro-3-(trifluoromethyl)benzaldehyde
Formula: C₈H₄ClF₃O
MW: 208.57
Melting Pt: 37…43 °C
Storage Temperature: Ambient
MDL Number: MFCD00792414
CAS Number: 93118-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR