6-Fluoro-L-tryptophan 97%

Supplier: Apollo Scientific

Synonyms: H-Trp(6-F)-OH, (S)-2-Amino-3-(6-fluoro-1H-indol-3-yl)propanoic acid

PC530036-1G PC530036-250MG
APOSPC530036-1GEA 317 EUR
APOSPC530036-1G APOSPC530036-250MG
6-Fluoro-L-tryptophan 97%
6-Fluoro-L-tryptophan
Formula: C₁₁H₁₁FN₂O₂
MW: 222.216 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00137731
CAS Number: 19310-00-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR