2-Fluoro-3-iodonitrobenzene

Supplier: Apollo Scientific
Warning

Synonyms: 2-Fluoro-1-iodo-3-nitrobenzene

PC53450-1G PC53450-250MG PC53450-5G
APOSPC53450-1GEA 493 EUR
APOSPC53450-1G APOSPC53450-250MG APOSPC53450-5G
2-Fluoro-3-iodonitrobenzene
2-Fluoro-3-iodonitrobenzene
Formula: C₆H₃FINO₂
MW: 266.996 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18396988
CAS Number: 1261782-23-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR