2-Amino-5-chloro-3-fluorobenzoic acid

Supplier: Apollo Scientific
Warning

PC53542-1G PC53542-250MG
APOSPC53542-1GEA 202 EUR
APOSPC53542-1G APOSPC53542-250MG
2-Amino-5-chloro-3-fluorobenzoic acid
2-Amino-5-chloro-3-fluorobenzoic acid
Formula: C₇H₅ClFNO₂
MW: 189.572 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11193641
CAS Number: 1028757-83-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR