4-(Trifluoromethyl)pyrimidine-2-carbonitrile ≥97%

Supplier: Apollo Scientific

PC540004-250MG
APOSPC540004-250MGEA 165 EUR
APOSPC540004-250MG
4-(Trifluoromethyl)pyrimidine-2-carbonitrile ≥97%
4-(Trifluoromethyl)pyrimidine-2-carbonitrile
Formula: C₆H₂F₃N₃
MW: 173.095 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD26386355
CAS Number: 1206459-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR