8-Chloro-6-fluorochroman-4-one ≥97%

Supplier: Apollo Scientific

PC540015-250MG
APOSPC540015-250MGEA 149 EUR
APOSPC540015-250MG
8-Chloro-6-fluorochroman-4-one ≥97%
8-Chloro-6-fluorochroman-4-one
Formula: C₉H₆ClFO₂
MW: 200.594 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11518446
CAS Number: 1092349-40-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR