Potassium-4-(trifluoroboratomethyl)thiomorpholine

Supplier: Apollo Scientific
Warning

Synonyms: Potassium trifluoro(thiomorpholinomethyl)borate

PC56184-1G PC56184-5G
APOSPC56184-1GEA 181 EUR
APOSPC56184-1G APOSPC56184-5G
Potassium-4-(trifluoroboratomethyl)thiomorpholine
Potassium 4-(trifluoroboratomethyl)thiomorpholine
Formula: C₅H₁₀BF₃KNS
MW: 223.11 g/mol
Storage Temperature: Freezer
MDL Number: MFCD10700162
CAS Number: 1150654-80-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR