4-Acetoxy-2,3,5,6-tetrafluorobenzoic acid

Supplier: Apollo Scientific
Warning

PC5714-250MG
APOSPC5714-250MGEA 21.9 EUR
APOSPC5714-250MG
4-Acetoxy-2,3,5,6-tetrafluorobenzoic acid
4-Acetoxy-2,3,5,6-tetrafluorobenzoic acid
Formula: C₉H₄F₄O₄
MW: 252.119 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03094301
CAS Number: 83789-90-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR