3-(1,1-Difluoroethyl)benzoic acid 95%

Supplier: Apollo Scientific
Warning

PC57526-250MG PC57526-1G PC57526-5G
APOSPC57526-250MGEA 190 EUR
APOSPC57526-250MG APOSPC57526-1G APOSPC57526-5G
3-(1,1-Difluoroethyl)benzoic acid 95%
3-(1,1-Difluoroethyl)benzoic acid
Formula: C₉H₈F₂O₂
MW: 186.16 g/mol
MDL Number: MFCD20640021
CAS Number: 55805-17-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR