5-Bromo-2-(difluoromethyl)benzonitrile ≥95%

Supplier: Apollo Scientific

PC57762-250MG PC57762-1G
APOSPC57762-250MGEA 261 EUR
APOSPC57762-250MG APOSPC57762-1G
5-Bromo-2-(difluoromethyl)benzonitrile ≥95%
5-Bromo-2-(difluoromethyl)benzonitrile
Formula: C₈H₄BrF₂N
MW: 232.02486 g/mol
MDL Number: MFCD18391066
CAS Number: 1128269-99-7
UN: 3439

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR