4-Fluoro-3-methylbenzylamine, technical

Supplier: Apollo Scientific

Synonyms: (4-Fluoro-3-methylphenyl)methanamine

PC57806-5G PC57806-25G PC57806-100G
APOSPC57806-5GEA 21.9 EUR
APOSPC57806-5G APOSPC57806-25G APOSPC57806-100G
4-Fluoro-3-methylbenzylamine, technical
4-Fluoro-3-methylbenzylamine
Formula: C₈H₁₀FN
MW: 139.17 g/mol
MDL Number: MFCD01631490
CAS Number: 261951-68-2
UN: 2735

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR