Novec HFE-72DE Engineered Fluid (3M)

Supplier: Apollo Scientific
Warning

PC6473-5KG
APOSPC6473-5KGEA 609 EUR
APOSPC6473-5KG
Novec HFE-72DE Engineered Fluid (3M)
Novec HFE-72DE Engineered Fluid

Cleaning and coating solvent

Boiling Pt: 45 °C
Density: 1.28 g/cm³ (25 °C)
Storage Temperature: Ambient
CAS Number: 839682-75-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR