1-Iodo-2-(heptafluoropropyl)cyclohexane (E/Z)

Supplier: Apollo Scientific

Synonyms: cis/trans-1-(Heptafluoropropyl)-2-iodocyclohexane

PC6544-250MG
APOSPC6544-250MGEA 170 EUR
APOSPC6544-250MG
1-Iodo-2-(heptafluoropropyl)cyclohexane (E/Z)
1-Iodo-2-(heptafluoropropyl)cyclohexane (E/Z)
Formula: C₉H₁₀F₇I
MW: 378.07 g/mol
Boiling Pt: 48…50 °C/1.3mmHg
Density: 1.8 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD02183508
CAS Number: 2728-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR