1,1,2-Trichloro-2,3,3-trifluorocyclobutane 97%

Supplier: Apollo Scientific
Warning

PC7060-5G
APOSPC7060-5GEA 137 EUR
APOSPC7060-5G
1,1,2-Trichloro-2,3,3-trifluorocyclobutane 97%
1,1,2-Trichloro-2,3,3-trifluorocyclobutane
Formula: C₄H₂Cl₃F₃
MW: 213.41 g/mol
Boiling Pt: 120 °C
Density: 1.594 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00039439
CAS Number: 697-17-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR