2,4,6-Trifluoroaniline 98%

Supplier: Apollo Scientific
Danger

PC7260-10G PC7260-250G PC7260-50G
APOSPC7260-10GEA 43.5 EUR
APOSPC7260-10G APOSPC7260-250G APOSPC7260-50G
2,4,6-Trifluoroaniline 98%
2,4,6-Trifluoroaniline
Formula: C₆H₄F₃N
MW: 147.1 g/mol
Boiling Pt: 57 °C/22mmHg
Melting Pt: 33…37 °C
Flash Pt: 58 °C
Storage Temperature: Ambient
MDL Number: MFCD00007650
CAS Number: 363-81-5
EINECS: 206-660-8
UN: 1325
ADR: 4.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR