2,3,4-Trifluorobenzyl alcohol 97%

Supplier: Apollo Scientific
Warning

PC7285L-25G PC7285L-5G
APOSPC7285L-25GEA 115 EUR
APOSPC7285L-25G APOSPC7285L-5G
2,3,4-Trifluorobenzyl alcohol 97%
2,3,4-Trifluorobenzyl alcohol
Formula: C₇H₅F₃O
MW: 162.11 g/mol
Melting Pt: 29.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00083551
CAS Number: 144284-24-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR