2-Deoxy-2-fluoro-D-glucose

Supplier: Apollo Scientific
Warning

Synonyms: 2-Fluoro-2-deoxy-D-glucose, 2-Deoxy-2-fluoro-D-glucopyranose, FDG

PC7306-100MG PC7306-25MG
APOSPC7306-100MGEA 488 EUR
APOSPC7306-100MG APOSPC7306-25MG
2-Deoxy-2-fluoro-D-glucose
2-Deoxy-2-fluoro-D-glucose

A substrate of hexokinase isozymes.

Formula: C₆H₁₁FO₅
MW: 182.15 g/mol
Melting Pt: 170…176 °C
Storage Temperature: Freezer
MDL Number: MFCD00077527
CAS Number: 29702-43-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR