4-(Trifluoromethoxy)benzoic acid 97%

Supplier: Apollo Scientific
Warning

PC7438-100G PC7438-25G
APOSPC7438-100GEA 72.5 EUR
APOSPC7438-100G APOSPC7438-25G
4-(Trifluoromethoxy)benzoic acid 97%
4-(Trifluoromethoxy)benzoic acid
Formula: C₈H₅F₃O₃
MW: 206.12 g/mol
Boiling Pt: 272
Melting Pt: 150…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00002541
CAS Number: 330-12-1
EINECS: 206-352-3

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR