3-(2-fluorophenyl)propan-1-ol 96%

Supplier: Apollo Scientific

PC900075-1G PC900075-250MG
APOSPC900075-1GEA 632 EUR
APOSPC900075-1G APOSPC900075-250MG
3-(2-fluorophenyl)propan-1-ol 96%
3-(2-fluorophenyl)propan-1-ol
Formula: C₉H₁₁FO
MW: 154.181 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09028696
CAS Number: 76727-24-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR