3-Fluoro-5-nitroanisole 97%

Supplier: Apollo Scientific

Synonyms: 1-Fluoro-3-methoxy-5-nitrobenzene

PC902146-10G PC902146-1G PC902146-5G
APOSPC902146-10GEA 690 EUR
APOSPC902146-10G APOSPC902146-1G APOSPC902146-5G
3-Fluoro-5-nitroanisole 97%
3-Fluoro-5-nitroanisole
Formula: C₇H₆FNO₃
MW: 171.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11848484
CAS Number: 7087-60-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR