4-Fluoro-2-methylthiobenzothiazole 98%

Supplier: Apollo Scientific

PC902199-1G PC902199-5G
APOSPC902199-1GEA 168 EUR
APOSPC902199-1G APOSPC902199-5G
4-Fluoro-2-methylthiobenzothiazole 98%
4-Fluoro-2-methylthiobenzothiazole
Formula: C₈H₆FNS₂
MW: 199.268 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12756452
CAS Number: 154327-25-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR