8-Fluoroquinoline-2-carbaldehyde 95%

Supplier: Apollo Scientific

PC905560-1G PC905560-250MG
APOSPC905560-1GEA 391 EUR
APOSPC905560-1G APOSPC905560-250MG
8-Fluoroquinoline-2-carbaldehyde 95%
8-Fluoroquinoline-2-carbaldehyde
Formula: C₁₀H₆FNO
MW: 175.159 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06824210
CAS Number: 904369-10-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR