2-(2-Fluorophenyl)-2-methylpropan-1-ol 95%

Supplier: Apollo Scientific

PC907490-1G
APOSPC907490-1GEA 647 EUR
APOSPC907490-1G
2-(2-Fluorophenyl)-2-methylpropan-1-ol 95%
2-(2-Fluorophenyl)-2-methylpropan-1-ol
Formula: C₁₀H₁₃FO
MW: 168.208 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11934574
CAS Number: 1177192-02-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR