2',6'-Difluoroacetanilide

Supplier: Apollo Scientific

Synonyms: N-(2,6-Difluorophenyl)acetamide

PC9976-1G PC9976-5G
APOSPC9976-1GEA 43.9 EUR
APOSPC9976-1G APOSPC9976-5G
2',6'-Difluoroacetanilide
2',6'-Difluoroacetanilide
Formula: C₈H₇F₂NO
MW: 171.144 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00119485
CAS Number: 3896-29-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR