Sulphuric acid 75% analysis_for
Supplier: PanReac AppliChem
Danger
123374.1212
APPC123374.1212EA 74.7 EUR
APPC123374.1212
Sulphuric acid 75% analysis_for
Sulphuric acid
Formula: H₂SO₄
MW: 98.08 g/mol
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.84
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 1830
ADR: 8 II
Order Now
Specification Test Results
Assay Min. 75%
Chloride Max. 0.00001%
Nitrate Max. 0.00002%
Phosphate Max. 0.00005%
Al Max. 0.000005%
As Max. 0.000001%
Ca Max. 0.00005%
Fe Max. 0.00002%
NA Max. 0.00005%
Ni Max. 0.000002%
PB Max. 0.000002%
Zn Max. 0.000005%