Dimethyl sulphoxide ≥99.9% Reag. Ph. Eur., ACS

Supplier: PanReac AppliChem

Synonyms: Methanesulphinylmethane, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO

131954.1611 131954.1612
APPC131954.1611EA 113 EUR
APPC131954.1611 APPC131954.1612
Dimethyl sulphoxide ≥99.9% Reag. Ph. Eur., ACS
Dimethyl sulphoxide
Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189
Melting Pt: 18.5
Flash Pt: 87
MDL Number: MFCD00002089
CAS Number: 67-68-5
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear, colourless liquid
Assay (G.C.)  Min. 99,9%
Identity IR Passes test
Acidity 0,001 mEq/g
Insoluble matter in H₂O Passes test
Non-volatile matter   0,001%
Methanol (G.C.) 0,05%
Dimethyl Sulfone (G.C.) 0,1%
Water (H₂O) 0,1%
Silver (Ag), by ICP 0,05 ppm
Aluminium (Al), by ICP 0,5 ppm
Arsenic (As), by ICP 0,05 ppm
Aurum (Au), by ICP 0,05 ppm
Boron (B), by ICP 0,02 ppm
Barium (Ba), by ICP 0,1 ppm
Berrylium (Be), by ICP 0,02 ppm
Bismuth (Bi), by ICP 0,05 ppm
Calcium (Ca), by ICP 0,5 ppm
Cadmium (Cd), by ICP 0,05 ppm
Cobalt (Co), by ICP 0,02 ppm
Chromium (Cr), by ICP 0,02 ppm
Copper (Cu), by ICP 0,02 ppm
Iron (Fe), by ICP 0,1 ppm
Gallium (Ga), by ICP 0,02 ppm
Germanium (Ge), by ICP 0,05 ppm
Mercury (Hg), by ICP 0,05 ppm
Indium (In), by ICP 0,05 ppm
Potassium (K), by ICP 0,1 ppm
Lithium (Li), by ICP 0,05 ppm
Magnesium (M)g, by ICP 0,1 ppm
Manganese (Mn), by ICP 0,02 ppm
Molybdenum (Mo), by ICP 0,02 ppm
Sodium (Na), by ICP 0,5 ppm
Nickel (Ni), by ICP 0,02 ppm
Phosphorous (P), by ICP 0,2 ppm
Lead (Pb), by ICP 0,1 ppm
Platinum (Pt), by ICP 0,02 ppm
Sulphur (S), by ICP 0,2 ppm
Antimony (Sb), by ICP 0,02 ppm
Silicon (Si), by ICP 0,2 ppm
Tin (Sn), by ICP 0,1 ppm
Strontium (Sr), by ICP 0,2 ppm
Titanium (Ti), by ICP 0,02 ppm
Thallium (Tl), by ICP 0,02 ppm
Vanadium (V), by ICP 0,02 ppm
Zinc (Zn), by ICP 0,1 ppm
Zirconium (Zr), by ICP 0,02 ppm
Solubility Miscible with water, hygroscopic
Vapour pressure (20 °C) 0,6 hPa
Viscosity 20 °C 2 mPa.s
Dipole moment 20 °C 3,9 Debye
Dielectric constant 20 °C 4,7


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR