Water, super gradient UPLC_UHPLC_suitable

Supplier: PanReac AppliChem

Synonyms: Hydrogen oxide

Nitrogen_filled

221074.1612
APPC221074.1612EA 59.1 EUR
APPC221074.1612
Water, super gradient UPLC_UHPLC_suitable
Water
Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 1.000 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Non-volatile Matter 0.0001%
Specific Conductance (25 °C) 1.0×10⁻⁶ ohm⁻¹ cm⁻¹
Suitability for Gradient According to ACS Passes Test
Gradient at 210 nm 5 mUA
Gradient at 254 nm 0.5 mUA
Fluorescence at 254 nm (as Quinine) 1 ppb
Fluorescence at 365 nm (as Quinine) 0.5 ppb
Transmittance at 200 nm (UV Spectrum, 1 cm Cell) ≥98%
Transmittance at 210 nm (UV Spectrum, 1 cm Cell) ≥98%
Transmittance at 254 nm (UV Spectrum, 1 cm Cell) ≥99%
Transmittance at 300 - 450 nm (UV Spectrum, 1 cm Cell) ≥99%

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR