Acetonitrile 99,9 GC_ASSAY_METHOD ACS, super gradient UHPLC

Supplier: PanReac AppliChem
Danger

Synonyms: Methyl cyanide, Ethyl nitrile, Cyanomethane, Ethanenitrile, Ethanoic acid nitrile

Filter 0,2 um_Inert_Gas_filled

221881.1611 221881.1612 221881.16153
APPC221881.1611EA 127 EUR
APPC221881.1611 APPC221881.1612 APPC221881.16153
Acetonitrile 99,9 GC_ASSAY_METHOD ACS, super gradient UHPLC
Acetonitrile
Formula: H₃CCN
MW: 41.05 g/mol
Boiling Pt: 80…82 °C
Melting Pt: –48 °C
Density: 0.781 g/cm³
Flash Pt: 2
Storage Temperature: Ambient
MDL Number: MFCD00001878
CAS Number: 75-05-8
EINECS: 200-835-2
UN: 1648
ADR: 3 III
Merck Index: 13,00071
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cef8be50fcca82050c5bda827ad9c5f57f4c5e27&vid=b80b5c39e654fcf6904b18b58e22a816e8e138b7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Minimum Assay (G.C.) 5 - 10
Identity IR Passes Test
Suitability for Gradient acc. to ACS Passes Test
APHA Colour Max. 10
Acidity Max. 0.0005 meq/g
Alkalinity Max. 0.0001 meq/g
Non-Volatile Matter Max. 0.0001%
Base Line Grift (210 nm) Max. 10 mUA
Water (H₂O) Max. 0.015%
Gradient at 210 nm Max. 1 mUA
Gradient at 254 nm Max. 0.5 mUA
Fluorescence at 254 nm (as Quinine) Max. 1 ppb
Fluorescence at 365 nm (as Quinine) Max. 0.5 ppb
Transmittance at 190 (Cut off) nm ≥30%
Transmittance at 193 nm ≥60%
Transmittance at 195 nm ≥80%
Transmittance at 200 nm ≥90%
Transmittance at 230 - 400 nm ≥98%
Rohrschneider Polarity (HPLC) Max. 5.8
Eluotropic Value E° (Al₂O₃ - HPLC) Max. 0.65


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR