Anti-BRD2 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
ARP34212_P050
AVIVARP34212_P050EA 401 EUR
AVIVARP34212_P050
Anti-BRD2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BRD2 (bromodomain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR