Anti-HSFX1 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
ARP35906_P050
AVIVARP35906_P050EA 401 EUR
AVIVARP35906_P050
Anti-HSFX1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HSFX1 (heat shock transcription factor family, X linked 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol HSFX1
Antigen name Heat shock transcription factor family, X linked 1
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 51402
Isotype
Western blot Yes
Epitope middle
Cross adsorption none
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR