Anti-SEPN1 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
ARP47656_P050
AVIVARP47656_P050EA 401 EUR
AVIVARP47656_P050
Anti-SEPN1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: SEPN1 (selenoprotein N, 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol SEPN1
Environmentally Preferable
Antigen name selenoprotein N, 1
Antigen synonyms MDRS1|RSMD1|SELN|CFTD|RSS
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity
Host Rabbit
Gene ID 57190
Isotype
Western blot Yes
Epitope C-terminal
Cross adsorption none
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR