Anti-ZNF320 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
ARP50901_P050
AVIVARP50901_P050EA 401 EUR
AVIVARP50901_P050
Anti-ZNF320 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ZNF320 (zinc finger protein 320)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR