Anti-C1QTNF2 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
ARP52422_P050
AVIVARP52422_P050EA 401 EUR
AVIVARP52422_P050
Anti-C1QTNF2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: C1QTNF2 (C1q and tumor necrosis factor related protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol C1QTNF2
Environmentally Preferable
Antigen name C1q and tumor necrosis factor related protein 2
Antigen synonyms 1810033K05Rik|CTRP2
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity
Host Rabbit
Gene ID 69183
Isotype
Western blot Yes
Epitope middle
Cross adsorption none
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR