Anti-KNG1 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
ARP59203_P050
AVIVARP59203_P050EA 401 EUR
AVIVARP59203_P050
Anti-KNG1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: KNG1 (kininogen 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR