Anti-TACC2 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
ARP61899_P050
AVIVARP61899_P050EA 401 EUR
AVIVARP61899_P050
Anti-TACC2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: TACC2 (transforming, acidic coiled-coil containing protein 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity:
Order Now

SPECIFICATIONS

Antigen symbol TACC2
Environmentally Preferable
Antigen name transforming, acidic coiled-coil containing protein 2
Antigen synonyms mKIAA4180
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity
Host Rabbit
Gene ID 57752
Isotype
Western blot Yes
Epitope C-terminal
Cross adsorption none
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR