Anti-MS4A3 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
ARP62012_P050
AVIVARP62012_P050EA 401 EUR
AVIVARP62012_P050
Anti-MS4A3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MS4A3 (membrane-spanning 4-domains, subfamily A, member 3 (hematopoietic cell-specific))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR