Anti-C17orf105 Rabbit Polyclonal Antibody

Supplier: Aviva Systems Biology
ARP69584_P050
AVIVARP69584_P050EA 401 EUR
AVIVARP69584_P050
Anti-C17orf105 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: C17orf105 (chromosome 17 open reading frame 105)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

SPECIFICATIONS

Antigen symbol C17orf105
Antigen name Chromosome 17 open reading frame 105
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Gene ID 284067
Isotype
Western blot Yes
Epitope middle
Cross adsorption none
Storage temperature Cool
Shipping temperature Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR