(2-Bromoethoxy)benzene 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: β-Bromophenetole, 2-Phenoxyethyl bromide, 2-Bromoethyl phenyl ether, ω-Bromophenetole

B21087.22 B21087.14 B21087.36
B21087.22EA 142 EUR
B21087.22 B21087.14 B21087.36
(2-Bromoethoxy)benzene 98
(2-Bromoethoxy)benzene

Purity: 98% CAS No.: 589-10-6 Molecular Formula: C6H5O(CH2)2Br Formula Weight: 201.07 Flash Point: 65°(149°F) Melting Point: 31-35° Boiling Point: 135-137°/20mm Beilstein Reference: 508290 MDL No.: MFCD00000234

Formula: BrCH₂CH₂OC₆H₅
MW: 201.06 g/mol
Boiling Pt: 135…137 °C (20 mmHg)
Melting Pt: 31…35 °C
Density: 1.35
Flash Pt: 65 °C (149 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000234
CAS Number: 589-10-6
EINECS: 209-634-4

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR