1-Bromo-4-fluoro-3-nitrobenzene 98

Supplier: Thermo Fisher Scientific
Danger

Synonyms: 4-Bromo-1-fluoro-2-nitrobenzene, 5-Bromo-2-fluoronitrobenzene

B22523.06 B22523.14
B22523.06EA 66.7 EUR
B22523.06 B22523.14
1-Bromo-4-fluoro-3-nitrobenzene 98
1-Bromo-4-fluoro-3-nitrobenzene

Purity: 98% CAS No.: 364-73-8 Molecular Formula: C6H3BrFNO2 Formula Weight: 220.01 Flash Point: 110°(230°F) Refractive Index: 1.5768 Melting Point: 18-19° Boiling Point: 240-241° Beilstein Reference: 2329084 UN No.: 2810 MDL No.: MFCD00129165

Formula: C₆H₃BrFNO₂
MW: 220 g/mol
Boiling Pt: 240…241 °C
Melting Pt: 18…19 °C
Density: 1.786
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00129165
CAS Number: 364-73-8
UN: 2810
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR