5-Methoxyisatin ≥98%

Supplier: Thermo Scientific
Warning

Synonyms: 5-Methoxy-1H-indole-2,3-dione, 5-Methoxyindoline-2,3-dione

B22742.06
B22742.06EA 56.5 EUR
B22742.06
5-Methoxyisatin ≥98%
5-Methoxyisatin

Purity: 98% CAS No.: 39755-95-8 Molecular Formula: C9H7NO3 Formula Weight: 177.16 Melting Point: 199-202° MDL No.: MFCD00169023

Formula: C₉H₇NO₃
MW: 177.16 g/mol
Melting Pt: 199…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00169023
CAS Number: 39755-95-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR