3,5-Diamino-1H-1,2,4-triazole ≥98%

Supplier: Thermo Fisher Scientific

Synonyms: 3,5-Diamino-1,2,4-triazole, Guanazole

B22775.06 B22775.14 B22775.22
B22775.06EA 35 EUR
B22775.06 B22775.14 B22775.22
3,5-Diamino-1H-1,2,4-triazole ≥98%
3,5-Diamino-1H-1,2,4-triazole

Purity: 98+% CAS No.: 1455-77-2 Molecular Formula: C2H5N5 Formula Weight: 99.10 Melting Point: ca 205° dec. Beilstein Reference: 112467

Formula: C₂H₅N₅
MW: 99.1 g/mol
Boiling Pt: 425
Melting Pt: ∼ 205 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00005233
CAS Number: 1455-77-2
EINECS: 215-937-2

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR