6-Fluoroindole 98

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 6-Fluoro-1H-indole

B24633.03 B24633.06 B24633.14
B24633.03EA 67.1 EUR
B24633.03 B24633.06 B24633.14
6-Fluoroindole 98
6-Fluoroindole

Purity: 98% CAS No.: 399-51-9 Molecular Formula: C8H6FN Formula Weight: 135.14 Melting Point: 74-77° Beilstein Reference: 112192

Formula: C₈H₆FN
MW: 135.14 g/mol
Melting Pt: 74…77 °C
Storage Temperature: Ambient
MDL Number: MFCD00056933
CAS Number: 399-51-9

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR