3-Nitro-o-xylene 99

Supplier: Thermo Fisher Scientific
Warning

Synonyms: 1,2-Dimethyl-3-nitrobenzene, 2,3-Dimethylnitrobenzene, 3-Nitro-o-xylene

B24917.22 B24917.36
B24917.22EA 39.9 EUR
B24917.22 B24917.36
3-Nitro-o-xylene 99
3-Nitro-o-xylene

Purity: 99% CAS No.: 83-41-0 Molecular Formula: C8H9NO2 Formula Weight: 151.17 Flash Point: 107°(224°F) Refractive Index: 1.5440 Melting Point: 13-15° Boiling Point: 244-246° Beilstein Reference: 2045105 UN No.: 1665 MDL No.: MFCD00007162

Formula: C₈H₉NO₂
MW: 151.16 g/mol
Boiling Pt: 244…246 °C
Melting Pt: 13…15 °C
Density: 1.129
Flash Pt: 107 °C (224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00007162
CAS Number: 83-41-0
EINECS: 201-474-3
UN: 1665
ADR: 6.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR