Thermo Scientific™ Basic Vortex Mixer
Supplier: Thermo Fisher Scientific
88882012
BARN88882012EA 366 EUR
BARN88882012
Thermo Scientific™ Basic Vortex Mixer
Shakers Vortex Shakers
The Basic Vortex Mixer provides comprehensive mixing capabilities for enzymatic and RIA assays, general test tube mixing, biochemical assays, viral dilution preparation, precipitated assays, cell suspension vortexing and tissue sample mixing.
  • Digital Speed display
  • Continuous and Touch modes
  • Convenient Speed adjustments by knob control
  • Stable for long-term operations
  • Multiple accessories for different vessels